3-Oxo-3-(1-pyrrolidinyl)propanenitrile ≥98%

Supplier: Thermo Scientific Chemicals
Warning

A12523-03 A12523-06
CAAAAA12523-03EA 47.24 CAD
CAAAAA12523-03 CAAAA12523-06 CAAAA12523-14
3-Oxo-3-(1-pyrrolidinyl)propanenitrile ≥98%
3-Oxo-3-(1-pyrrolidinyl)propanenitrile

Purity: 98+% CAS No.: 14227-95-3 Molecular Formula: C7H10N2O Formula Weight: 138.17 Odor: Odorless Melting Point: 71-74° Beilstein Reference: 637151 MDL No.: MFCD00020838

Formula: C₇H₁₀N₂O
MW: 138.17 g/mol
Melting Pt: 71…74 °C
Storage Temperature: Ambient
MDL Number: MFCD00020838
CAS Number: 14227-95-3
EINECS: 238-096-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR