Methyl-α-D-glucopyranoside 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
α-D-Methylglucoside, (2R,3S,4S,5R,6S)-2-(Hydroxymethyl)-6-methoxytetrahydro-2H-pyran-3,4,5-triol
A12484-22
A12484-36
CAAAA12484-22EA
61.46
CAD
CAAAA12484-22
CAAAA12484-36
Methyl-α-D-glucopyranoside 98%
Methyl α-D-glucopyranoside
Soluble in water. Insoluble in ether
Purity: 98+%
CAS No.: 97-30-3
Molecular Formula: C7H14O6
Formula Weight: 194.18
Melting Point: 166-171°
Solubility: Soluble in water. Insoluble in ether
Beilstein Reference: 81568
Merck Reference: 14,6080
Optical Rotation: +159° (c=10 in water)
MDL No.: MFCD00064086
Formula:
C₇H₁₄O₆ MW: 194.18 g/mol Melting Pt: 166…171 °C Density: 1.460 Storage Temperature: Ambient |
MDL Number:
MFCD00064086 CAS Number: 97-30-3 EINECS: 202-571-3 Merck Index: 14,6080 |
Specification Test Results
Optical rotation | +159° (c=10 in water) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...