Diethyl diallylmalonate ≥97%

Supplier: Thermo Scientific Chemicals
Warning

A12460-30 A12460-09 A12460-18
CAAAAA12460-30EA 1608.44 CAD
CAAAAA12460-30 CAAAA12460-09 CAAAA12460-18
Diethyl diallylmalonate ≥97%
Diethyl diallylmalonate

Purity: 98% CAS No.: 3195-24-2 Molecular Formula: C13H20O4 Formula Weight: 240.30 Flash Point: 110°(230°F) Refractive Index: 1.4460 Boiling Point: 242-244° Beilstein Reference: 1790529 MDL No.: MFCD00009126

Formula: C₁₃H₂₀O₄
Boiling Pt: 242…244 °C
Density: 0.993
Flash Pt: >110 °C(230 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009126
CAS Number: 3195-24-2
EINECS: 221-696-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR