3-Phenyl-1,2,4-triazole-5-thiol hydrate 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12427-03
A12427-06
CAAAA12427-03EA
45.19
CAD
CAAAA12427-03
CAAAA12427-06
3-Phenyl-1,2,4-triazole-5-thiol hydrate 98%
3-Phenyl-1,2,4-triazole-5-thiol hydrate
Purity: 98%
CAS No.: 3414-94-6
Molecular Formula: C8H7N3S.xH2O
Formula Weight: 177.23(anhy)
Melting Point: 253-258°
Beilstein Reference: 135556
MDL No.: MFCD00150547
Formula:
C₈H₇N₃S.xH₂O Melting Pt: 253…258 °C Storage Temperature: Ambient |
MDL Number:
MFCD00150547 CAS Number: 3414-94-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...