Ethyl-4-pentenoate ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12422-03
A12422-22
CAAAAA12422-03EA
50.88
CAD
CAAAAA12422-03
CAAAA12422-22
Ethyl-4-pentenoate ≥98%
Ethyl-4-pentenoate
Purity: 98+%
CAS No.: 1968-40-7
Molecular Formula: CH2=CHCH2CH2CO2CH2CH3
Formula Weight: 128.17
Flash Point: 37°(98°F)
Refractive Index: 1.4170
Boiling Point: 146°
Beilstein Reference: 1751643
UN No.: 3272
MDL No.: MFCD00036562
Formula:
C₇H₁₂O₂ Boiling Pt: 146 °C Density: 0.900 Flash Pt: 37 °C (98 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00036562 CAS Number: 1968-40-7 EINECS: 217-818-0 UN: 3272 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...