4-Isopropylphenylhydrazine hydrochloride 98%

Supplier: Thermo Scientific Chemicals
Warning

A12416-03 A12416-06
CAAAA12416-03EA 53.4 CAD
CAAAA12416-03 CAAAA12416-06
4-Isopropylphenylhydrazine hydrochloride 98%
4-Isopropylphenylhydrazine hydrochloride

Purity: 98% CAS No.: 118427-29-5 Molecular Formula: C9H14N2.HCl Formula Weight: 186.69 Melting Point: 203° dec. MDL No.: MFCD00013461

Formula: C₉H₁₅ClN₂
Melting Pt: 203 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00013461
CAS Number: 118427-29-5
UN: 2811
ADR: 6.1,III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR