L-Histidine methyl ester dihydrochloride ≥98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: L(+)-Histidine methyl ester dihydrochloride, H-His-OMe.2HCl, L-Methyl histidinate dihydrochloride, H-His-OMe·2HCl

A12386-06 A12386-14 A12386-22
CAAAA12386-06EA 44.71 CAD
CAAAA12386-06 CAAAA12386-14 CAAAA12386-22
L-Histidine methyl ester dihydrochloride ≥98%
L-Histidine methyl ester dihydrochloride

Purity: 98+% CAS No.: 7389-87-9 Molecular Formula: C7H11N3O2.2HCl Formula Weight: 242.11 Storage Temperature: 2°C to 8°C Melting Point: 198° dec. Beilstein Reference: 3572010 Optical Rotation: +10° (c=2 in water) MDL No.: MFCD00012701

Formula: C₇H₁₁N₃O₂
MW: 242.10 g/mol
Melting Pt: 198 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00012701
CAS Number: 7389-87-9
EINECS: 230-973-9

Order Now

Specification Test Results

Optical rotation +10° (c=2 in water)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR