Sulfadiazine 99%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
N-(2-Pyrimidinyl)sulphanilamide, N-(2-Pyrimidinyl)sulfanilamide, 4-Amino-N-(pyrimidin-2-yl)benzenesulfonamide, 4-Amino-N-(pyrimidin-2-yl)benzenesulphonamide, 4-Amino-N-(2-pyrimidinyl)benzenesulfonamide, 4-Amino-N-(2-pyrimidinyl)benzenesulphonamide
A12370-0B
A12370-18
A12370-30
CAAAA12370-0BEA
475.58
CAD
CAAAA12370-0B
CAAAA12370-18
CAAAA12370-30
Sulfadiazine 99%
Sulfadiazine
Purity: 99%
CAS No.: 68-35-9
Molecular Formula: C10H10N4O2S
Formula Weight: 250.28
Physical Form: Powder
Appearance: White
Melting Point: 253-256°
MDL No.: MFCD00006065
Formula:
C₁₀H₁₀N₄O₂S MW: 250.28 g/mol Melting Pt: 253…256 °C Storage Temperature: Ambient |
MDL Number:
MFCD00006065 CAS Number: 68-35-9 EINECS: 200-685-8 Merck Index: 14,8903 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...