L-serine methyl ester hydrochloride 98%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Ser-OMe.HCl

A12355-06 A12355-14 A12355-22
CAAAA12355-06EA 63.2 CAD
CAAAA12355-06 CAAAA12355-14 CAAAA12355-22
L-serine methyl ester hydrochloride 98%
L-serine methyl ester hydrochloride

Purity: 98% CAS No.: 5680-80-8 Molecular Formula: C4H9NO3.HCl Formula Weight: 155.58 Storage Temperature: -30°C to -10°C Melting Point: 155° dec. Beilstein Reference: 3559591 Optical Rotation: +5° (c=2 in methanol) MDL No.: MFCD00063680

Formula: C₄H₉NO₃
MW: 155.58 g/mol
Melting Pt: 155 °C (dec.)
Storage Temperature: Freezer
MDL Number: MFCD00063680
CAS Number: 5680-80-8
EINECS: 227-140-7

Order Now

Specification Test Results

Optical rotation +5° (c=2 in methanol)


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR