Sodium glycolate 97%
Supplier: Thermo Scientific Chemicals
A12341-0E
A12341-22
A12341-36
CAAAA12341-0EEA
891.12
CAD
CAAAA12341-0E
CAAAA12341-22
CAAAA12341-36
Sodium glycolate 97%
Sodium glycolate
Purity: 97%
CAS No.: 2836-32-0
Molecular Formula: C2H3NaO3
Formula Weight: 98.03
Physical Form: Lumpy powder
Appearance: White
Melting Point: 210-218°
MDL No.: MFCD00065358
Formula:
C₂H₃NaO₃ MW: 98.03 g/mol Melting Pt: 210…218 °C Storage Temperature: Ambient |
MDL Number:
MFCD00065358 CAS Number: 2836-32-0 EINECS: 220-624-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...