Ethyl 3-Methylpyrazole-5-carboxylate 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12304-14
A12304-06
CAAAAA12304-14EA
922.72
CAD
CAAAAA12304-14
CAAAA12304-03
CAAAA12304-06
Ethyl 3-Methylpyrazole-5-carboxylate 97%
Ethyl 3-Methylpyrazole-5-carboxylate
Purity: 97%
CAS No.: 4027-57-0
Molecular Formula: C7H10N2O2
Formula Weight: 154.17
Melting Point: 80-83°
Beilstein Reference: 4192
MDL No.: MFCD00052514
Formula:
C₇H₁₀N₂O₂ Melting Pt: 80…83 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052514 CAS Number: 1226862-09-4 EINECS: 223-702-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...