Ethyl thiophene-2-carboxylate 98+%
Supplier: Thermo Scientific Chemicals
Warning
|
A12302-09
A12302-18
CAAAAA12302-09EA
44.71
CAD
CAAAAA12302-09
CAAAAA12302-18
Ethyl thiophene-2-carboxylate 98+%
Ethyl thiophene-2-carboxylate
Purity: 98+%
CAS No.: 2810-04-0
Molecular Formula: C7H8O2S
Formula Weight: 156.20
Flash Point: 88°(190°F)
Refractive Index: 1.5260
Boiling Point: 217-218°
Beilstein Reference: 112678
MDL No.: MFCD00005436
Formula:
C₇H₈O₂S Boiling Pt: 217…218 °C Density: 1,161 Flash Pt: 88 °C(190 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00005436 CAS Number: 2810-04-0 EINECS: 220-554-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...