Dichloromethylenedimethyliminium chloride 95%

Supplier: Thermo Scientific Chemicals
Danger

A12300-14 A12300-22
CAAAA12300-14EA 170.64 CAD
CAAAA12300-14 CAAAA12300-22
Dichloromethylenedimethyliminium chloride 95%
Dichloromethylenedimethyliminium chloride

Fieser: 4,135 5,195 6,170 8,156 13,106 20,154


Purity: 95% CAS No.: 33842-02-3 Molecular Formula: C3H6Cl3N Formula Weight: 162.45 Storage Temperature: 2°C to 8°C Melting Point: ca 185° dec. Beilstein Reference: 4157987 UN No.: 3261 MDL No.: MFCD00011870

Formula: C₃H₆Cl₃N
Melting Pt: ca 185 °C (dec.)
Storage Temperature: Refrigerator
MDL Number: MFCD00011870
CAS Number: 33842-02-3
EINECS: 251-695-4
UN: 3261
ADR: "8",III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR