1-Benzothiophene-2-carboxamide 97%
Supplier: Thermo Scientific Chemicals
A12292-03
A12292-06
CAAAAA12292-14EA
1636.47
CAD
CAAAAA12292-14
CAAAA12292-03
CAAAA12292-06
1-Benzothiophene-2-carboxamide 97%
1-Benzothiophene-2-carboxamide
Purity: 97%
CAS No.: 6314-42-7
Molecular Formula: C9H7NOS
Formula Weight: 177.23
Melting Point: 176-178°
Beilstein Reference: 4467
MDL No.: MFCD00052505
Formula:
C₉H₇NOS Melting Pt: 176…178 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052505 CAS Number: 6314-42-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...