Methyl-3,5-dimethylbenzoate 98%

Supplier: Thermo Scientific Chemicals

Synonyms: Methyl 3,5-dimethylbenzoate

A12287-06 A12287-14
CAAAA12287-06EA 72.36 CAD
CAAAA12287-06 CAAAA12287-14
Methyl-3,5-dimethylbenzoate 98%
Methyl 3,5-dimethylbenzoate

Purity: 98% CAS No.: 25081-39-4 Molecular Formula: C10H12O2 Formula Weight: 164.21 Flash Point: 106°(224°F) Melting Point: 31-33° Boiling Point: 239-240° MDL No.: MFCD00077971

Formula: C₁₀H₁₂O₂
MW: 164.21 g/mol
Boiling Pt: 239…240 °C
Melting Pt: 31…33 °C
Density: 1,027
Flash Pt: 106 °C(224 °F)
Storage Temperature: Ambient
MDL Number: MFCD00077971
CAS Number: 25081-39-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR