2-Chloro-N-(hydroxymethyl)acetamide 98%

Supplier: Thermo Scientific Chemicals
Danger

A12247-09 A12247-18
CAAAA12247-09EA 126.08 CAD
CAAAA12247-09 CAAAA12247-18
2-Chloro-N-(hydroxymethyl)acetamide 98%
2-Chloro-N-(hydroxymethyl)acetamide

Purity: 98% CAS No.: 2832-19-1 Molecular Formula: C3H6ClNO2 Formula Weight: 123.54 Storage Temperature: 2°C to 8°C Melting Point: 97-103° Beilstein Reference: 1811765 UN No.: 3261 MDL No.: MFCD00021961

Formula: C₃H₆ClNO₂
MW: 123.54 g/mol
Melting Pt: 97…103 °C
Storage Temperature: Refrigerator
MDL Number: MFCD00021961
CAS Number: 2832-19-1
EINECS: 220-598-9
UN: 2923
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR