4-(4-Nitrophenylazo)resorcinol 90+%

Supplier: Thermo Scientific Chemicals
Warning

A12232-14 A12232-22 A12232-36
CAAAA12232-14EA 80.26 CAD
CAAAA12232-14 CAAAA12232-22 CAAAA12232-36
4-(4-Nitrophenylazo)resorcinol 90+%
4-(4-Nitrophenylazo)resorcinol

Purity: 95% CAS No.: 74-39-5 Molecular Formula: C12H9N3O4 Formula Weight: 259.22 Melting Point: ca 185° dec. Beilstein Reference: 674709 Merck Reference: 14,5695 MDL No.: MFCD00007310

Formula: C₁₂H₉N₃O₄
Melting Pt: ca 185 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00007310
CAS Number: 74-39-5
EINECS: 200-808-5
Merck Index: 14,5695

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR