2-Amino-3'-nitroacetophenone hydrochloride 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12205-03
A12205-06
CAAAA12205-03EA
106.81
CAD
CAAAA12205-03
CAAAA12205-06
2-Amino-3'-nitroacetophenone hydrochloride 98%
2-Amino-3'-nitroacetophenone hydrochloride
Purity: 98%
CAS No.: 36765-84-1
Molecular Formula: C8H8N2O3.HCl
Formula Weight: 216.62
Melting Point: 210-212° dec.
UN No.: 1759
MDL No.: MFCD00051997
Formula:
C₈H₉ClN₂O₃ Melting Pt: 210…212 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00051997 CAS Number: 36765-84-1 UN: 1759 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...