Sulfur trioxide pyridine complex 98% active SO₃ ∼ 48-50%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12202-09
A12202-0B
A12202-18
A12202-30
CAAAAA12202-09EA
44.71
CAD
CAAAAA12202-09
CAAAA12202-0B
CAAAA12202-18
CAAAA12202-30
Sulfur trioxide pyridine complex 98% active SO₃ ∼ 48-50%
Sulphur trioxide pyridine complex
Fieser: 1,1127 2,393 3,275 4,473
Purity: 98%
CAS No.: 26412-87-3
Molecular Formula: C5H5NO3S
Formula Weight: 159.16
Melting Point: ca 155-165°
Beilstein Reference: 3704116
UN No.: 3261
MDL No.: MFCD00012437
Formula:
C₅H₅NO₃S MW: 159.16 g/mol Melting Pt: ∼ 155…165 °C Storage Temperature: Ambient |
MDL Number:
MFCD00012437 CAS Number: 26412-87-3 EINECS: 247-683-3 UN: 3261 ADR: 8,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...