6-Mercaptopurine monohydrate 98%

Supplier: Thermo Scientific Chemicals
Warning

A12197-03 A12197-06 A12197-14
CAAAA12197-03EA 42.03 CAD
CAAAA12197-03 CAAAA12197-06 CAAAA12197-14
6-Mercaptopurine monohydrate 98%
6-Mercaptopurine monohydrate

Purity: 98% CAS No.: 6112-76-1 Molecular Formula: C5H4N4S.H2O Formula Weight: 170.19 (152.17anhy) Melting Point: ca 315° dec. Beilstein Reference: 4012091 MDL No.: MFCD03854445

Formula: C₅H₄N₄S·H₂O
Melting Pt: ∼ 315 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD03854445
CAS Number: 6112-76-1
EINECS: 200-037-4
Merck Index: 14,5871

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR