N,N'-Diallyl-L-tartardiamide 99%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A12195-18
A12195-30
CAAAA12195-18EA
103.02
CAD
CAAAA12195-18
CAAAA12195-30
N,N'-Diallyl-L-tartardiamide 99%
N,N'-Diallyl-L-tartardiamide
Applications: Cross-linking co-monomer for the polymerization of soluble polyacrylamide gels
Purity: 99%
CAS No.: 58477-85-3
Molecular Formula: C10H16N2O4
Formula Weight: 228.25
Melting Point: 184-186°
Beilstein Reference: 1712934
Formula:
C₁₀H₁₆N₂O₄ MW: 228.25 g/mol Melting Pt: 184…186 °C Storage Temperature: Ambient |
MDL Number:
MFCD00008640 CAS Number: 58477-85-3 EINECS: 261-277-3 |
Specification Test Results
Optical rotation | +118° (c=3 in methanol) |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...