tert-Butyldimethylsilyl trifluoromethanesulfonate 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12174-06
A12174-14
CAAAA12174-06EA
120.24
CAD
CAAAA12174-06
CAAAA12174-14
tert-Butyldimethylsilyl trifluoromethanesulfonate 98%
tert-Butyldimethylsilyl trifluoromethanesulphonate
Fieser: 10,63 12,86 13,50 15,54 17,55 19,51
Purity: 98%
CAS No.: 69739-34-0
Molecular Formula: C7H15F3O3SSi
Formula Weight: 264.34
Storage Temperature: -30°C to -10°C
Flash Point: 36°(96°F)
Refractive Index: 1.3860
Boiling Point: 65-67°/12mm
Beilstein Reference: 2370068
UN No.: 2920
MDL No.: MFCD00000405
Formula:
C₇H₁₅F₃O₃SSi Boiling Pt: 65…67 °C/12 mm Density: 1.151 Flash Pt: 36 °C(96 °F) Storage Temperature: Freezer |
MDL Number:
MFCD00000405 CAS Number: 69739-34-0 EINECS: 274-102-0 UN: 2920 ADR: "8",II |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...