Methyl dimethoxyacetate 97+%
Supplier: Thermo Scientific Chemicals
Warning
|
A12171-09
A12171-18
A12171-30
CAAAAA12171-06EA
98.73
CAD
CAAAAA12171-06
CAAAA12171-09
CAAAA12171-18
CAAAA12171-30
Methyl dimethoxyacetate 97+%
Methyl dimethoxyacetate
Purity: 97+%
CAS No.: 89-91-8
Molecular Formula: (CH3O)2CHCO2CH3
Formula Weight: 134.13
Flash Point: 63°(145°F)
Refractive Index: 1.4050
Boiling Point: 160-163°
Beilstein Reference: 1757582
MDL No.: MFCD00008484
Formula:
C₅H₁₀O₄ Boiling Pt: 160…163 °C Density: 1.098 Flash Pt: 63 °C(145 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008484 CAS Number: 89-91-8 EINECS: 201-950-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...