Penta-O-acetyl-β-D-galactopyranose 98%

Supplier: Thermo Scientific Chemicals

Synonyms: β-D-Galactose Pentaacetate, (2S,3R,4S,5S,6R)-6-(Acetoxymethyl)tetrahydro-2H-pyran-2,3,4,5-tetrayl tetraacetate

A12163-14 A12163-22
CAAAA12163-06EA 86.16 CAD
CAAAA12163-06 CAAAA12163-14 CAAAA12163-22
Penta-O-acetyl-β-D-galactopyranose 98%
Penta-O-acetyl-beta-D-galactopyranose

Purity: 98% CAS No.: 4163-60-4 Molecular Formula: C16H22O11 Formula Weight: 390.34 Melting Point: 142-146° Beilstein Reference: 98853 Optical Rotation: +24° (c=4 in chloroform) MDL No.: MFCD00063259

Formula: C₁₆H₂₂O₁₁
MW: 390.34 g/mol
Melting Pt: 142…146 °C
Storage Temperature: Ambient
MDL Number: MFCD00063259
CAS Number: 4163-60-4
EINECS: 224-008-0

Order Now

Specification Test Results

Optical rotation +24° (c=4 in chloroform)

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR