Glycine hydrochloride (H-Gly-OH.HCl) 98%

Supplier: Thermo Scientific Chemicals

Synonyms: H-Gly-OH·HCl, Glycine hydrochloride

A12162-0E A12162-22 A12162-36
CAAAA12162-0EEA 1336.68 CAD
CAAAA12162-0E CAAAA12162-22 CAAAA12162-36
Glycine hydrochloride (H-Gly-OH.HCl) 98%
Glycine hydrochloride (H-Gly-OH.HCl)

Purity: 98% CAS No.: 6000-43-7 Molecular Formula: H2NCH2CO2H.HCl Formula Weight: 111.53 Odor: Odorless Melting Point: 175-178° Beilstein Reference: 3909617 Merck Reference: 14,4491 MDL No.: MFCD00012872

Formula: C₂H₆ClNO₂
MW: 111.53 g/mol
Melting Pt: 175…178 °C
Storage Temperature: Ambient
MDL Number: MFCD00012872
CAS Number: 6000-43-7
EINECS: 227-841-8
Merck Index: 14,4491

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR