trans-Indole-3-acrylic acid ≥98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
trans-3-Indoleacrylic acid
A12143-03
A12143-06
CAAAA12143-03EA
81.84
CAD
CAAAA12143-03
CAAAA12143-06
CAAAA12143-14
trans-Indole-3-acrylic acid ≥98%
trans-Indole-3-acrylic acid
Purity: 98+%
CAS No.: 29953-71-7
Molecular Formula: C11H9NO2
Formula Weight: 187.20
Storage Temperature: 2°C to 8°C
Melting Point: ca 185°dec.
Beilstein Reference: 6317
MDL No.: MFCD00005633
Formula:
C₁₁H₉NO₂ MW: 187.20 g/mol Melting Pt: ca 185 °C (dec.) Storage Temperature: Refrigerator |
MDL Number:
MFCD00005633 CAS Number: 29953-71-7 EINECS: 214-872-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...