5-Phenyl-1,3-cyclohexanedione 97%
Supplier: Thermo Scientific Chemicals
A12136-06
A12136-14
CAAAA12136-06EA
76.63
CAD
CAAAA12136-06
CAAAA12136-14
5-Phenyl-1,3-cyclohexanedione 97%
5-Phenyl-1,3-cyclohexanedione
Purity: 97%
CAS No.: 493-72-1
Molecular Formula: C12H12O2
Formula Weight: 188.23
Melting Point: ca 185° dec.
Beilstein Reference: 1103470
Formula:
C₁₂H₁₂O₂ Melting Pt: ca 185 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00051846 CAS Number: 493-72-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...