Ethyl-4-oxo-3-piperidinecarboxylate hydrochloride 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: Ethyl 4-Oxo-3-piperidinecarboxylate HCl, 4-Oxo-3-piperidinecarboxylic Acid Ethyl Ester Hydrochloride, 3-Ethoxycarbonyl-4-piperidone Hydrochloride, Ethyl 4-piperidone-3-carboxylate hydrochloride, 3-Carbethoxy-4-piperidone hydrochloride, Ethyl 4-Oxo-3-piperidinecarboxylate Hydrochloride

A12131-06 A12131-14
CAAAAA12131-06EA 96.22 CAD
CAAAAA12131-06 CAAAAA12131-14 CAAAAA12131-22
Ethyl-4-oxo-3-piperidinecarboxylate hydrochloride 97%
Ethyl-4-oxo-3-piperidinecarboxylate hydrochloride

Purity: 97% CAS No.: 4644-61-5 Molecular Formula: C8H13NO3.HCl Formula Weight: 207.66 Melting Point: ca 172° dec. Beilstein Reference: 3711279 MDL No.: MFCD00043267

Formula: C₈H₁₃NO₃
MW: 207.65 g/mol
Melting Pt: ca 172 °C (dec.)
Storage Temperature: Ambient
MDL Number: MFCD00043267
CAS Number: 4644-61-5
EINECS: 225-073-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR