Dowex® 1X2 (Cl), Dowex® 50-100 mesh

Supplier: Thermo Scientific Chemicals

A12130-0B A12130-18 A12130-30
CAAAA12130-0BEA 887.96 CAD
CAAAA12130-0B CAAAA12130-18 CAAAA12130-30
Dowex® 1X2 (Cl), Dowex® 50-100 mesh
Dowex® 1X2 (Cl)

Fieser: 1,511 4,267 6,302 7,182 8,263


CAS No.: 9085-42-1 Odor: Amine-like MDL No.: MFCD00132715

Storage Temperature: Ambient
MDL Number: MFCD00132715
CAS Number: 9085-42-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR