trans,trans-Dibenzylideneacetone 98+%

Supplier: Thermo Scientific Chemicals

Synonyms: (1E,4E)-1,5-Diphenylpenta-1,4-dien-3-one

A12095-06 A12095-14 A12095-22
CAAAAA12095-06EA 48.82 CAD
CAAAAA12095-06 CAAAA12095-14 CAAAA12095-22
trans,trans-Dibenzylideneacetone 98+%
trans,trans-Dibenzylideneacetone

Purity: 98+% CAS No.: 35225-79-7 Molecular Formula: C17H14O Formula Weight: 234.30 Melting Point: 112-114° Beilstein Reference: 608434 Merck Reference: 14,3006 MDL No.: MFCD00004790

Formula: C₁₇H₁₄O
Melting Pt: 112…114 °C
Storage Temperature: Ambient
MDL Number: MFCD00004790
CAS Number: 35225-79-7
Merck Index: 14,3006

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR