α-D-Glucose-1-phosphate disodium salt tetrahydrate 98%
Supplier: Thermo Scientific Chemicals
A12064-06
A12064-14
CAAAA12064-06EA
284.4
CAD
CAAAA12064-06
CAAAA12064-14
α-D-Glucose-1-phosphate disodium salt tetrahydrate 98%
α-D-Glucose-1-phosphate disodium salt tetrahydrate
Purity: 98%
CAS No.: 56401-20-8
Molecular Formula: C6H11Na2O9P.4H2O
Formula Weight: 376.16 (304.10anhy)
Storage Temperature: -30°C to -10°C
Melting Point: ca 138°
MDL No.: MFCD00150839
Formula:
C₆H₁₁O₉PNa₂·4H₂O Melting Pt: ∼ 138 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00150839 CAS Number: 150399-99-8 EINECS: 260-154-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...