Di-2-thienyl ketone 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2,2'-Carbonyldi(thiophene), Di(thiophen-2-yl)methanone
A12062-03
A12062-06
CAAAAA12062-14EA
1416.64
CAD
CAAAAA12062-14
CAAAA12062-03
CAAAA12062-06
Di-2-thienyl ketone 98%
Di-2-thienyl ketone
Purity: 98%
CAS No.: 704-38-1
Molecular Formula: C9H6OS2
Formula Weight: 194.28
Melting Point: 87-90°
Beilstein Reference: 129168
MDL No.: MFCD00052096
Formula:
C₉H₆OS₂ Melting Pt: 87…90 °C Storage Temperature: Ambient |
MDL Number:
MFCD00052096 CAS Number: 704-38-1 UN: 2811 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...