Ethyl-3-hydroxyhexanoate 97%

Supplier: Thermo Scientific Chemicals

Synonyms: Ethyl 3-hydroxyhexanoate

A12060-14 A12060-22
CAAAA12060-14EA 90.06 CAD
CAAAA12060-14 CAAAA12060-22
Ethyl-3-hydroxyhexanoate 97%
Ethyl 3-hydroxyhexanoate

Purity: 97% CAS No.: 2305-25-1 Molecular Formula: C8H16O3 Formula Weight: 160.21 Flash Point: 94°(202°F) Refractive Index: 1.4298 Boiling Point: 90-92°/14mm MDL No.: MFCD00036604

Formula: C₈H₁₆O₃
MW: 160.21 g/mol
Boiling Pt: 90…92 °C/14 mm
Density: 0,974
Flash Pt: 94 °C(202 °F)
Storage Temperature: Ambient
MDL Number: MFCD00036604
CAS Number: 2305-25-1
EINECS: 218-973-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR