trans-Cinnamoyl chloride 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12016-0E
A12016-22
A12016-36
CAAAA12016-0EEA
1540.5
CAD
CAAAA12016-0E
CAAAA12016-22
CAAAA12016-36
trans-Cinnamoyl chloride 97%
trans-Cinnamoyl chloride
Purity: 97%
CAS No.: 17082-09-6
Molecular Formula: C9H7ClO
Formula Weight: 166.61
Flash Point:
Formula:
C₉H₇ClO Boiling Pt: 256…258 °C Melting Pt: 35…37 °C Density: 1.162 Flash Pt: >110 °C(230 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00000732 CAS Number: 17082-09-6 EINECS: 203-065-5 UN: 3261 ADR: "8",III Merck Index: 14,2301 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...