Pivalonitrile ≥98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A12011-14
A12011-22
CAAAAA12011-14EA
136.83
CAD
CAAAAA12011-14
CAAAAA12011-22
Pivalonitrile ≥98%
Pivalonitrile
Purity: 98+%
CAS No.: 630-18-2
Molecular Formula: (CH3)3CCN
Formula Weight: 83.13
Flash Point: 4°(39°F)
Odor: Odorless
Melting Point: 16-18°
Boiling Point: 105-106°
UN No.: 3273
MDL No.: MFCD00001847
Formula:
C₅H₉N Boiling Pt: 105…106 °C Melting Pt: 16…18 °C Density: 0.757 Flash Pt: 4 °C (39 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00001847 CAS Number: 630-18-2 EINECS: 211-133-0 UN: 1992 ADR: 3,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...