L-Leucine-p-nitroanilide 99%
Supplier: Thermo Scientific Chemicals
A11994-03
A11994-06
CAAAA11994-03EA
85.95
CAD
CAAAA11994-03
CAAAA11994-06
L-Leucine-p-nitroanilide 99%
L-Leucine-p-nitroanilide
Purity: 99%
CAS No.: 4178-93-2
Molecular Formula: C12H17N3O3
Formula Weight: 251.29
Storage Temperature: 2°C to 8°C
Melting Point: 89-90°
Beilstein Reference: 2218433
MDL No.: MFCD00063681
Formula:
C₁₂H₁₇N₃O₃ Melting Pt: 89…90 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00063681 CAS Number: 4178-93-2 EINECS: 224-047-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...