N-(Cyanoacetyl)morpholine 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
3-Morpholino-3-oxopropanenitrile, 3-(Morpholin-4-yl)-3-oxopropanenitrile
A11969-03
A11969-06
CAAAAA11969-03EA
45.19
CAD
CAAAAA11969-03
CAAAA11969-06
CAAAA11969-14
N-(Cyanoacetyl)morpholine 98%
N-(Cyanoacetyl)morpholine
Purity: 98%
CAS No.: 15029-32-0
Molecular Formula: C7H10N2O2
Formula Weight: 154.17
Odor: Odorless
Melting Point: 85-88°
Beilstein Reference: 135525
MDL No.: MFCD00023353
Formula:
C₇H₁₀N₂O₂ MW: 154.17 g/mol Melting Pt: 85…88 °C Storage Temperature: Ambient |
MDL Number:
MFCD00023353 CAS Number: 15029-32-0 EINECS: 239-112-1 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...