m-Toluoylacetonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 3-Oxo-3-(m-tolyl)propanenitrile

A11965-03 A11965-06
CAAAA11965-03EA 45.35 CAD
CAAAA11965-03 CAAAA11965-06
m-Toluoylacetonitrile 97%
m-Toluoylacetonitrile

Purity: 97% CAS No.: 53882-81-8 Molecular Formula: C10H9NO Formula Weight: 159.19 Melting Point: 71-75° Beilstein Reference: 2717335 MDL No.: MFCD00067922

Formula: C₁₀H₉NO
MW: 159.19 g/mol
Melting Pt: 71…75 °C
Storage Temperature: Ambient
MDL Number: MFCD00067922
CAS Number: 53882-81-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR