N-Benzoyl-N-phenylhydroxylamine 98%
Supplier: Thermo Scientific Chemicals
Synonyms:
Tantalon, N-Phenylbenzohydroxamic Acid, N-Hydroxy-N-phenylbenzamide
A11950-06
A11950-14
CAAAA11950-06EA
127.35
CAD
CAAAA11950-06
CAAAA11950-14
N-Benzoyl-N-phenylhydroxylamine 98%
N-Benzoyl-N-phenylhydroxylamine
Purity: 98%
CAS No.: 304-88-1
Molecular Formula: C13H11NO2
Formula Weight: 213.24
Melting Point: 118-120°
Beilstein Reference: 2212449
MDL No.: MFCD00002111
Formula:
C₁₃H₁₁NO₂ MW: 213.24 g/mol Melting Pt: 118…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00002111 CAS Number: 304-88-1 EINECS: 206-158-9 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...