tert-Butyl ethyl malonate 95%

Supplier: Thermo Scientific Chemicals
Warning

A11927-14 A11927-22
CAAAA11927-06EA 138.06 CAD
CAAAA11927-06 CAAAA11927-14 CAAAA11927-22
tert-Butyl ethyl malonate 95%
tert-Butyl ethyl malonate

Purity: 95% CAS No.: 32864-38-3 Molecular Formula: C9H16O4 Formula Weight: 188.22 Flash Point: 87°(188°F) Refractive Index: 1.4160 Boiling Point: 98-100°/22mm Beilstein Reference: 1776681 MDL No.: MFCD00009193

Formula: C₉H₁₆O₄
MW: 188.22 g/mol
Boiling Pt: 98…100 °C/22 mm
Density: 0.994
Flash Pt: 87 °C(188 °F)
Storage Temperature: Ambient
MDL Number: MFCD00009193
CAS Number: 32864-38-3
EINECS: 251-269-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR