4-Methylesculetin 97%
Supplier: Thermo Scientific Chemicals
A11902-06
A11902-14
A11902-22
CAAAA11902-06EA
49.93
CAD
CAAAA11902-06
CAAAA11902-14
CAAAA11902-22
4-Methylesculetin 97%
4-Methylesculetin
Purity: 97%
CAS No.: 529-84-0
Molecular Formula: C10H8O4
Formula Weight: 192.17
Melting Point: ca 275° dec.
Beilstein Reference: 162550
MDL No.: MFCD00006859
Formula:
C₁₀H₈O₄ Melting Pt: ca 275 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00006859 CAS Number: 529-84-0 EINECS: 208-470-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...