Aminoiminomethanesulfinic acid 98%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11885-22
A11885-36
CAAAA11885-22EA
59.25
CAD
CAAAA11885-22
CAAAA11885-36
Aminoiminomethanesulfinic acid 98%
Aminoiminomethanesulfinic acid
Fieser: 4,506 5,668 6,586 8,232
Purity: 98%
CAS No.: 1758-73-2
Molecular Formula: CH4N2O2S
Formula Weight: 108.12
Storage Temperature: 2°C to 8°C
Melting Point: ca 126° dec.
Beilstein Reference: 506653
UN No.: 3341
MDL No.: MFCD00002397
Formula:
CH₄N₂O₂S Melting Pt: ca 126 °C (dec.) Density: 1.68 Storage Temperature: Refrigerator |
MDL Number:
MFCD00002397 CAS Number: 1758-73-2 EINECS: 217-157-8 UN: 3341 ADR: "4.2",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...