2-Hydroxy-1,4-naphthoquinone 98+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11880-09
A11880-18
A11880-30
CAAAA11880-09EA
42.34
CAD
CAAAA11880-09
CAAAA11880-18
CAAAA11880-30
2-Hydroxy-1,4-naphthoquinone 98+%
2-Hydroxy-1,4-naphthoquinone
Fieser: 1,484
Purity: 98+%
CAS No.: 83-72-7
Molecular Formula: C10H6O3
Formula Weight: 174.16
Melting Point: ca 193° dec.
Beilstein Reference: 1565260
Merck Reference: 14,5393
MDL No.: MFCD00001678
Formula:
C₁₀H₆O₃ Melting Pt: ca 193 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00001678 CAS Number: 83-72-7 EINECS: 201-496-3 Merck Index: 14,5393 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...