Methyl-3-(bromomethyl)benzoate 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
A11826-06
A11826-14
CAAAA11826-03EA
71.5
CAD
CAAAA11826-03
CAAAA11826-06
CAAAA11826-14
Methyl-3-(bromomethyl)benzoate 97%
Methyl 3-(bromomethyl)benzoate
Purity: 97%
CAS No.: 1129-28-8
Molecular Formula: C9H9BrO2
Formula Weight: 229.07
Melting Point: 43-45°
Boiling Point: 115°/0.8mm
UN No.: 3261
MDL No.: MFCD00051437
Formula:
C₉H₉BrO₂ Boiling Pt: 115 °C/0.8 mm Melting Pt: 43…45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00051437 CAS Number: 1129-28-8 EINECS: 419-100-4 UN: 3261 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...