1-(2-Cyanophenyl)piperidine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11784-03
A11784-06
CAAAA11784-03EA
22.75
CAD
CAAAA11784-03
CAAAA11784-06
1-(2-Cyanophenyl)piperidine 97%
1-(2-Cyanophenyl)piperidine
Purity: 97%
CAS No.: 72752-52-4
Molecular Formula: C12H14N2
Formula Weight: 186.26
Melting Point: 41-45°
Boiling Point: 122-125°/1mm
UN No.: 3077
MDL No.: MFCD00049221
Formula:
C₁₂H₁₄N₂ Boiling Pt: 122…125 °C/1 mm Melting Pt: 41…45 °C Storage Temperature: Ambient |
MDL Number:
MFCD00049221 CAS Number: 72752-52-4 EINECS: 427-330-1 UN: 3077 ADR: "9",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...