4-Bromo-3,5-dimethyl-1H-pyrazole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Bromo-3,5-dimethylpyrazole
A11759-22
A11759-06
A11759-14
CAAAAA11759-22EA
944.84
CAD
CAAAAA11759-22
CAAAA11759-06
CAAAA11759-14
4-Bromo-3,5-dimethyl-1H-pyrazole 98%
4-Bromo-3,5-dimethyl-1H-pyrazole
Purity: 98%
CAS No.: 3398-16-1
Molecular Formula: C5H7BrN2
Formula Weight: 175.03
Melting Point: 118-123°
Beilstein Reference: 108534
Formula:
C₅H₇BrN₂ MW: 175.03 g/mol Melting Pt: 118…123 °C Storage Temperature: Ambient |
MDL Number:
MFCD00005242 CAS Number: 3398-16-1 EINECS: 222-263-2 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...