4,6-Diphenyl-2-pyrone 98%
Supplier: Thermo Scientific Chemicals
A11755-03
A11755-06
CAAAA11755-03EA
78.37
CAD
CAAAA11755-03
CAAAA11755-06
4,6-Diphenyl-2-pyrone 98%
4,6-Diphenyl-2-pyrone
Purity: 98%
CAS No.: 17372-52-0
Molecular Formula: C17H12O2
Formula Weight: 248.27
Odor: Odorless
Melting Point: 135-138°
MDL No.: MFCD00031018
Formula:
C₁₇H₁₂O₂ Melting Pt: 135…138 °C Storage Temperature: Ambient |
MDL Number:
MFCD00031018 CAS Number: 17372-52-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...