5-Hydantoin acetic acid 98%
Supplier: Thermo Scientific Chemicals
A11731-06
A11731-14
CAAAAA11731-06EA
48.35
CAD
CAAAAA11731-06
CAAAAA11731-14
5-Hydantoin acetic acid 98%
5-Hydantoin acetic acid
Purity: 98%
CAS No.: 5427-26-9
Molecular Formula: C5H6N2O4
Formula Weight: 158.11
Melting Point: ca 217° dec.
Beilstein Reference: 83035
MDL No.: MFCD00005267
Formula:
C₅H₆N₂O₄ Melting Pt: ca 217 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00005267 CAS Number: 5427-26-9 EINECS: 226-574-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...