5-Chloro-1,3-dimethyl-1H-pyrazole 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 5-Chloro-1,3-dimethylpyrazole

A11730-06 A11730-14
CAAAA11730-06EA 164.32 CAD
CAAAA11730-06 CAAAA11730-14
5-Chloro-1,3-dimethyl-1H-pyrazole 98%
5-Chloro-1,3-dimethyl-1H-pyrazole

Purity: 98% CAS No.: 54454-10-3 Molecular Formula: C5H7ClN2 Formula Weight: 130.58 Refractive Index: 1.4860 Boiling Point: 153-155° MDL No.: MFCD00051650

Formula: C₅H₇ClN₂
MW: 130.58 g/mol
Boiling Pt: 153…155 °C
Density: 1.137
Storage Temperature: Ambient
MDL Number: MFCD00051650
CAS Number: 54454-10-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR