(Triphenylphosphoranylidene)acetaldehyde, (max. 3% H₂O) 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
A11709-14
A11709-22
A11709-09
CAAAAA11709-14EA
255.96
CAD
CAAAAA11709-14
CAAAAA11709-22
CAAAA11709-09
(Triphenylphosphoranylidene)acetaldehyde, (max. 3% H₂O) 97%
(Triphenylphosphoranylidene)acetaldehyde
Purity: 97%
CAS No.: 2136-75-6
Molecular Formula: (C6H5)3P=CHCHO
Formula Weight: 304.33
Storage Temperature: 2°C to 8°C
Melting Point: 183-188°
Beilstein Reference: 523797
MDL No.: MFCD00006994
Formula:
C₂₀H₁₇OP Melting Pt: 183…188 °C Storage Temperature: Refrigerator |
MDL Number:
MFCD00006994 CAS Number: 2136-75-6 EINECS: 218-375-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...